missing translation for 'onlineSavingsMsg'
Learn More

NuSieve™ 3:1 Agarose, Lonza™

Product Code. 11537785
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11537785 25 g Each
11547785 500 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11537785 Supplier Lonza Supplier No. 50091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easy-to-handle gels enhance the speed of gel processing and blotting. Lonza™ NuSieve™ 3:1 Agarose is a molecular biology grade, standard melting temperature agarose that yields strong gels for fine resolution of small DNA, RNA, and PCR products ≤ 1 kb.

  • Designed for analytical electrophoresis
  • Resolution of DNA fragments up to 1,000 bp, but is capable of resolving DNA fragments  from 10 bp up to 1,500 bp
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6

Specifications

Quantity 25 g
Grade Molecular Biology
Chemical Name or Material Agarose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.