missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ GelStar™ Nucleic Acid Gel Stain

Product Code. 11550767
Click to view available options
Quantity:
2 x 250 μL
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 11550767

Brand: Lonza™ 50535

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use to detect small quantities of DNA and RNA. GelStar™ Nucleic Acid Gel Stain exhibits exceptional signal-to-noise ratio with minimal background.

  • Detects as little as 20pg of DNA or 3ng of RNA
  • Exhibits exceptional signal-to-noise ratio with minimal background
  • Compatible with agarose or polyacrylamide gels
  • Can be added to gel before casting or used for post-electrophoresis staining
  • Can be visualized with a standard 300nm UV transilluminator or the Dark Reader™ transilluminator
  • Ideal alternative to silver staining
  • For research use only

Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Concentration 10,000X
Product Type DNA/RNA Stain
Stain Type Nucleic Acid Gel Stain
Form Liquid
For Use With (Application) Agarose or Polyacrylamide gels
Quantity 2 x 250 μL
Storage Requirements −20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.