missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System RNA Marker

Product Code. 11587805
Click to view available options
Quantity:
50 μL
Unit Size:
50 reactions
This item is not returnable. View return policy
Do not exceed 5μL per well load volume on FlashGel cassettes

Product Code. 11587805

Brand: Lonza™ 50577

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Do not exceed 5μL per well load volume on FlashGel cassettes

Consists of RNA transcripts 0.5 to 9kbp

Designed to optimize performance of the FlashGel System (Cat. No. BMA57067): optimized for the unique requirements of RNA. Provides consistent results in gene expression, Northern analysis, cDNA library construction and cDNA labeling for microarrays.

  • 50μL
  • Band sizes: 0.5/1/1.5/3/5/9kbp
  • Results in 30 minutes or less
  • Detect <10ng RNA per band
  • Avoid hazardous reagents and contaminating RNases
  • Cassettes contain precast, pre-stained agarose gels and buffer: no need for gel preparation, buffer addition or gel staining
  • Clean, enclosed system, provided ready-to-load
  • Denature FlashGel RNA markers with Formaldehyde sample Buffer (Cat. No. BW50571) for 5 minutes at 65°C (149°F)
  • For native RNA, combine with FlashGel loading dye (Cat. No. BMA50462)

  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals

  • Verification and analysis of total RNA
  • Quick checks of native RNA
  • Checking for RNA degradation and mRNA purity

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/mL
Product Type RNA Molecular Weight Marker/Ladder
Form Liquid
For Use With (Application) Accurate size estimation of DNA fragments
No. of Bands 6
Quantity 50 μL
Storage Requirements −80°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.