missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System Recovery Cassettes

Product Code. 11557865
Click to view available options
Separation Range:
10 to 1000 bp
50 to 10,000 bp
Unit Size:
Each
Pack of 9
This item is not returnable. View return policy

Product Code. 11557865

Brand: Lonza™ 57051

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Eliminates agarose gel preparation, band excision and purification. Lonza™ FlashGel™ - System Recovery Cassettes deliver recovery, free from inhibitors and UV-induced damage, in a simple 5-10 minute protocol.

  • Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
  • Recover samples directly, without band excision or purification
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY

Tekniske data

Concentration 1.2%
Product Type System Recovery Cassettes
Separation Range 50 to 10,000 bp
Wells 8 + 1-well
Formulation 1.2% Agarose
For Use With (Application) Subsequent re-amplification, cloning or other techniques
Kit Contents Agarose Gel, Stain and Buffer
Material Agarose
Quantity 9 Pack
Storage Requirements 18° to 26°C
Length (English) Cassette 4.5 in.
Length (English) Gel 2.7 in.
Length (Metric) Cassette 11.5 cm
Length (Metric) Gel 7 cm
Width (English) Cassette 4.2 in.
Width (Metric) Cassette 10.7 cm
Width (Metric) Gel 8.4 cm
Height (English) Cassette 0.6 in.
Height (Metric) Cassette 1.7 cm
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.