missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System DNA Markers

Product Code. p-4683829
Click to view available options
No. of Bands:
6
8
For Use With (Application):
1.2% FlashGel DNA Cassette
2.2% FlashGel DNA Cassette
Double-tier FlashGel DNA Cassette
Unit Size:
500µL
This item is not returnable. View return policy
Do not exceed 5μL per well load volume on FlashGel cassettes

Product Code. 11547805

Brand: Lonza™ 50473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Do not exceed 5μL per well load volume on FlashGel cassettes

Provide accurate size estimation of DNA fragments. FlashGel™ System DNA Markerswere designed to optimize performance of the FlashGel™ system and come ready-to-use.

  • Ready-to-load molecular weight standards
  • Separates DNA in real time: 2 to 7 minutes
  • Clean, sharp separation and straight, uniform sample lanes
  • Watch DNA migration as it happens: Monitor gel runs right at the bench, without UV light
  • Highly sensitive system allows a reduction in DNA levels, saving time and money
  • Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
  • Detect <0.1ng DNA or <10ng total RNA
  • Recommended for FlashGel DNA cassettes

 

FlashGel 1.2% DNA Marker

  • Sizes: 100/200/300/500/800/1250/2000/4000bp
  • Recommended for 1.2% FlashGel DNA cassettes
FlashGel 2.2% DNA Marker
  • Sizes: 50/100/150/200/300/500/800/1500bp
  • Recommended for 2.2% FlashGel DNA cassettes
FlashGel Double Tier DNA Marker
  • Sizes: 100/300/500/800/1500/3000bp
  • Recommended for double tier FlashGel DNA cassettes

 

 

  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Formulation 5μL Load 1:5 dilution
For Use With (Application) 1.2% FlashGel DNA Cassette
No. of Bands 8
Quantity 500 μL
Storage Requirements 4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.