Learn More
Lonza™ FlashGel™ RNA Marker Kit
Product Code. 11537855
Click to view available options
Quantity:
9 Pack
Unit Size:
Each
Product Code. 11537855
Brand: Lonza™ 57024
Currently Unavailable: We have no information when this product will be available.
View alternative products
Includes
FlashGel RNA Cassettes, 1.2% agarose (9 Pk.), 12 + 1 well single tier; Formaldehyde Sample Buffer; FlashGel RNA Marker; and AccuGENE™ Molecular Biology Water
Markers for fast analysis, recovery, and documentation of RNA. FlashGel™ RNA Marker Kit is recommended for verification and analysis of total RNA, quick checks of native RNA, and checking RNA degradation and mRNA purity.
  • Provide nearly instant results
  • Load samples, watch bands migrate and get data in as little as two minutes
  • Needs no gel preparation, band excision, purification, and UV light
  • Complete separation, recovery and documentation safely at the bench
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Analysis of total RNA, quick checks of native RNA and checking RNA degradation and mRNA purity
Kit Contents FlashGel™ RNA Cassettes 1.2% Agarose 12 + 1 well, Formaldehyde Sample Buffer, FlashGel™ RNA Marker and AccuGENE Molecular Biology Water
Quantity 9 Pack