missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ AccuGENE™ PBS
Description
AccuGENE Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
- High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted.
- Efficient - Ready-made solutions eliminate experiment preparation time.
- Flexible - Customized solutions are available to meet individual needs
Specifications
Specifications
| Concentration | 10X |
| For Use With (Application) | Ideal for a wide range of molecular biology applications |
| pH | 7.4 |
| Quantity | 1000mL |
| Formulation | KH2PO4 0.017M, Na2HPO4 0.05M, NaCl 1.5M |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?