missing translation for 'onlineSavingsMsg'
Learn More

AccuGENE™ 1X TE Buffer, Lonza™

Product Code. 12838281
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12838281 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12838281 Supplier Lonza Supplier No. 51235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A ready to use solution ideal for a wide range of molecular biology applications

AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1X TE Buffer
Concentration 1X
For Use With (Application) Molecular biology applications
pH 7.4
Quantity 500 mL
Formulation Tris, 0.001M EDTA (Disodium salt) 0.01M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.