missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ AccuGENE™ 1M Tris HCl Buffers
Click to view available options
pH:
7.2
7.4
8.0
Unit Size:
1 L
Each
Description
- Prepared with 18 megohm water
- Filtered using 0.2μ filter and filled into sterile bottles
- Manufactured according to strict quality control standards to ensure consistency
- Guaranteed DNase, RNase and protease-free
- Customized solutions are available to meet individual needs
Specifications
Specifications
| Concentration | 1 M |
| DNase | Yes |
| For Use With (Application) | Ideal for a wide range of molecular biology applications |
| pH | 7.4 |
| Quantity | 1000 mL |
For research or manufacturing use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction