missing translation for 'onlineSavingsMsg'
Learn More

LONRF3 Antibody (OTI9G7), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-44960-30ul

 View more versions of this product

Product Code. 30306361

  • £195.00 / 30µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LONRF3 Monoclonal antibody specifically detects LONRF3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LONRF3
Monoclonal
Unconjugated
PBS (pH 7.3) 1% BSA, 50% glycerol
Mouse
>95%
RUO
Primary
Human
Antibody
IgG1
Western Blot
OTI9G7
Western Blot 1:2000
LON peptidase N-terminal domain and ring finger 3, LON peptidase N-terminal domain and RING finger protein 3, MGC119463, MGC119465, RING finger protein 127FLJ22612, RNF127
Human recombinant protein fragment corresponding to amino acids 206-456 of human LONRF3 (NP_001027026) produced in E.coli.
30 μL
Zinc Finger
79836
Store at -20 degrees C. Avoid freeze/thaw cycles.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.