missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LONRF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566079
This item is not returnable.
View return policy
Description
Immunogen sequence: LAPDDNSLLLL RAELYLTMKN YEQALQDASA ACQNEPLLIK GHQVKAQALS GLGRSKEVLK EFLYC Highest antigen sequence identity to the following orthologs - mouse 56%, rat 82%.
LONRF2 contains one Lon domain, one RING type zinc finger and six TPR repeats. The function of LONRF2 is unknown. There are two named isoforms.
Specifications
| LONRF2 | |
| Polyclonal | |
| Unconjugated | |
| LONRF2 | |
| LON peptidase N-terminal domain and ring finger 2; LON peptidase N-terminal domain and RING finger protein 2; LONRF2; Neuroblastoma apoptosis-related protease; RING finger protein 192; RNF192 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 164832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q1L5Z9 | |
| LONRF2 | |
| Recombinant Human LONRF2. Recombinant protein control fragment (Product #RP-106771). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction