missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LONP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100020
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LONP1.
This gene encodes a mitochondrial matrix protein that belongs to the Lon family of ATP-dependent proteases. This protein mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides in the mitochondrial matrix. It may also have a chaperone function in the assembly of inner membrane protein complexes, and participate in the regulation of mitochondrial gene expression and maintenance of the integrity of the mitochondrial genome. Decreased expression of this gene has been noted in a patient with hereditary spastic paraplegia. Alternatively spliced transcript variants have been found for this gene.
Specifications
| LONP1 | |
| Polyclonal | |
| Unconjugated | |
| LONP1 | |
| 1200017E13Rik; CODASS; h LonHS; HAMAP-Rule:MF_03120}; hLON; hLON ATP-dependent protease; Lon; lon peptidase 1, mitochondrial; lon protease homolog, mitochondrial; lon protease homolog, mitochondrial {ECO:0000255; Lon protease-like protein; lon protease-like protein {ECO:0000255; LONHs; LONP; LONP {ECO:0000255; LONP1; Mitochondrial ATP-dependent protease Lon; mitochondrial ATP-dependent protease Lon {ECO:0000255; mitochondrial lon protease-like protein; PIM1; protease, serine, 15; Prss15; Serine protease 15; serine protease 15 {ECO:0000255 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170916, 74142, 9361 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36776, Q8CGK3, Q924S5 | |
| LONP1 | |
| A synthesized peptide derived from human LONP1(Accession P36776), corresponding to amino acid residues L825-Q875. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion