missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LONP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551692
This item is not returnable.
View return policy
Description
Immunogen sequence: VEEKIKQTHR KYLLQEQLKI IKKELGLEKD DKDAIEEKFR ERLKELVVPK HVMDVVDEEL SKLGLLDNHS SEFNVTRNYL DWLTSIPWGK YSNENLDLAR AQAVLEEDHY GMEDVKKRIL EFIAVSQLRG STQGKILCFY GP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
This gene encodes a mitochondrial matrix protein that belongs to the Lon family of ATP-dependent proteases. This protein mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides in the mitochondrial matrix. It may also have a chaperone function in the assembly of inner membrane protein complexes, and participate in the regulation of mitochondrial gene expression and maintenance of the integrity of the mitochondrial genome. Decreased expression of this gene has been noted in a patient with hereditary spastic paraplegia. Alternatively spliced transcript variants have been found for this gene.
Specifications
| LONP1 | |
| Polyclonal | |
| Unconjugated | |
| LONP1 | |
| 1200017E13Rik; CODASS; h LonHS; HAMAP-Rule:MF_03120}; hLON; hLON ATP-dependent protease; Lon; lon peptidase 1, mitochondrial; lon protease homolog, mitochondrial; lon protease homolog, mitochondrial {ECO:0000255; Lon protease-like protein; lon protease-like protein {ECO:0000255; LONHs; LONP; LONP {ECO:0000255; LONP1; Mitochondrial ATP-dependent protease Lon; mitochondrial ATP-dependent protease Lon {ECO:0000255; mitochondrial lon protease-like protein; PIM1; protease, serine, 15; Prss15; Serine protease 15; serine protease 15 {ECO:0000255 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170916, 74142, 9361 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.30 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P36776, Q8CGK3, Q924S5 | |
| LONP1 | |
| Recombinant protein corresponding to Human LONP1. Recombinant protein control fragment (Product #RP-99402). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction