Learn More
Invitrogen™ LONP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120887
Description
Positive test controls include: A-549, HL-60, LO2, HT-1080, BT-474, Mouse heart, Mouse kidney, Mouse liver, Rat kidney. The target is usually found in the following locations: Mitochondrion matrix. Immunogen sequence: VFPHLPLIAI TRNPVFPRFI KIIEVKNKKL VELLRRKVRL AQPYVGVFLK RDDSNESDVV ESLDEIYHTG TFAQIHEMQD LGDKLRMIVM GHRRVHISRQ LEVEPEEPEA ENKHKPRRKS KRGKKEAEDE LSARHPAELA MEPTPELPAE VLMVEVENVV HEDFQVTEEV KALTAEIVKT IRDIIALNPL YRESVLQMMQ AGQRVVDNPI YLSDMGAALT GAESHELQDV LEETNIPKRL YKALSLLKKE FELSKLQQRL GREVEEKIKQ THRKYLLQEQ L.
This gene encodes a mitochondrial matrix protein that belongs to the Lon family of ATP-dependent proteases. This protein mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides in the mitochondrial matrix. It may also have a chaperone function in the assembly of inner membrane protein complexes, and participate in the regulation of mitochondrial gene expression and maintenance of the integrity of the mitochondrial genome. Decreased expression of this gene has been noted in a patient with hereditary spastic paraplegia. Alternatively spliced transcript variants have been found for this gene.
Specifications
| LONP1 | |
| Polyclonal | |
| Unconjugated | |
| LONP1 | |
| 1200017E13Rik; CODASS; h LonHS; HAMAP-Rule:MF_03120}; hLON; hLON ATP-dependent protease; Lon; lon peptidase 1, mitochondrial; lon protease homolog, mitochondrial; lon protease homolog, mitochondrial {ECO:0000255; Lon protease-like protein; lon protease-like protein {ECO:0000255; LONHs; LONP; LONP {ECO:0000255; LONP1; Mitochondrial ATP-dependent protease Lon; mitochondrial ATP-dependent protease Lon {ECO:0000255; mitochondrial lon protease-like protein; PIM1; protease, serine, 15; Prss15; Serine protease 15; serine protease 15 {ECO:0000255 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170916, 74142, 9361 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36776, Q8CGK3, Q924S5 | |
| LONP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-400 of human LONP1 (NP_004784.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.