missing translation for 'onlineSavingsMsg'
Learn More

LONP1 Antibody (LON20/H1), DyLight 550, Novus Biologicals™

Product Code. 18125259 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18125259 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18125259 Supplier Novus Biologicals Supplier No. NBP250478R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LONP1 Monoclonal specifically detects LONP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LONP1
Applications Western Blot, ELISA, Immunohistochemistry, KnockDown
Classification Monoclonal
Clone LON20/H1
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.53, hLON, hLON ATP-dependent protease, LON, lon peptidase 1, mitochondrial, lon protease homolog, mitochondrial, Lon protease-like protein, LONHs, LONP, Mitochondrial ATP-dependent protease Lon, mitochondrial lon peptidase 1, mitochondrial lon protease-like protein, PIM1, protease, serine, 15, PRSS15MGC1498, Serine protease 15
Gene Symbols LONP1
Host Species Mouse
Immunogen KLH-peptide conjugate - N-CEKDDKDAIEEKFRERLKE-C
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Epigenetics, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9361
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.