missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Longdaysin, MedChemExpress
Description
Longdaysin is a inhibitor of the Wnt/β-catenin signaling pathway, which exerts antitumor effect through blocking CK1δ/ε-dependent Wnt signaling. Longdaysin inhibits CK1α, CK1δ, CDK7, and ERK2 with IC50s of 5.6 μM, 8.8 μM, 29 μM, and 52 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Longdaysin |
| CAS | 1353867-91-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H16F3N5 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 125 mg/mL (372.77 mM; Need ultrasonic) |
| SMILES | FC(F)(F)C1=CC=CC(CNC2=NC=NC3=C2N=CN3C(C)C)=C1 |
| Molecular Weight (g/mol) | 335.33 |
| Formula Weight | 335.33 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?