missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Long-span starter bay chipboard shelf, 2500mm x 2700mm x 600mm
Description
Heavy duty shelving that is ideal for hand loaded storage. Quick, easy boltless assembly - frames and beams simply clip together. Pre-welded frames - no assembly required. Level heights adjust every 62. 5mm. Beams in grey and upright frames in blue.
- Number of shelves: 3
- Dimensions (H x W x D): 2500mm x 2700mm x 600mm
- Shelf capacity UDL: 400kg
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction