Learn More
Manutan Long-span shelving steel panel, 2000mm x 2700mm x 1000mm
Description
Heavy duty shelving that is ideal for hand loaded storage. Quick, easy boltless assembly - frames and beams simply clip together. Pre-welded frames - no assembly required. With beams in grey and upright frames in blue. Level heights adjust every 62. 5mm. Shelving loads of up to 740kg UDL.
- Number of shelves: 3
- Dimensions (H x W x D): 2000mm x 2700mm x 1000mm
- Shelf capacity UDL: 740kg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.