Learn More
Hamilton™ Long Life Instrument Syringes
Long life syringes were developed for extreme conditions that include harsh chemicals as well as abrasive particulate matter.
Brand:  Hamilton™ 8250-22
Code : NEW
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 10698025
  • £210.00 / Each
Estimated Shipment: 18-06-2024
to see stock.
Description
Description
  • Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
  • Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications
Specifications
Borosilicate Glass, PTFE, UHMWPE
Instrument Syringe
100 μL
7.73 mm
1/Ea.
Luer Lock