missing translation for 'onlineSavingsMsg'
Learn More

Lonafarnib, MedChemExpress

Product Code. 16496318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496318 10 mg 10mg
16458907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496318 Supplier Medchem Express Supplier No. HY15136
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lonafarnib (Sch66336) is a potent and orally active farnesyl transferase (FTase) inhibitor. Lonafarnib inhibits the activities of H-ras, K-ras and N-ras with IC50 values of 1.9 nM, 5.2 nM and 2.8 nM, respectively. Lonafarnib also has anti-hepatitis delta virus (HDV) activities.

Lonafarnib (Sch66336) is a potent and orally active farnesyl transferase (FTase) inhibitor. Lonafarnib inhibits the activities of H-ras, K-ras and N-ras with IC50 values of 1.9 nM, 5.2 nM and 2.8 nM, respectively. Lonafarnib also has anti-hepatitis delta virus (HDV) activities.

Specifications

Chemical Name or Material Lonafarnib
CAS 193275-84-2
Color White
Purity Grade Notes Research
Molecular Formula C27H31Br2ClN4O2
Quantity 10 mg
Synonym Sch66336
Solubility Information DMSO : 50 mg/mL (78.27 mM; Need ultrasonic)
SMILES O=C(N1CCC(CC(N2CCC([C@@H]3C4=C(Br)C=C(Cl)C=C4CCC5=CC(Br)=CN=C53)CC2)=O)CC1)N
Molecular Weight (g/mol) 638.82
Formula Weight 638.82
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.