missing translation for 'onlineSavingsMsg'
Learn More

Lomustine, MedChemExpress

Product Code. 16441096
Encompass_Preferred
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
This item is not returnable. View return policy

Product Code. 16441096

Brand: Medchem Express HY13669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lomustine (CCNU; NSC 79037) is a DNA alkylating agent, with antitumor activity.

Lomustine (CCNU

Specifications

Chemical Name or Material Lomustine
Physical Form Solid
CAS 13010-47-4
Color Light Yellow
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C9H16ClN3O2
For Use With (Application) Cancer-programmed cell death
Synonym CCNU NSC 79037
Solubility Information DMSO : ≥ 100 mg/mL (427.90 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1CCCCC1)N(CCCl)N=O
Molecular Weight (g/mol) 233.7
Formula Weight 233.7
Percent Purity 98.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.