missing translation for 'onlineSavingsMsg'
Learn More

Lomustine, MedChemExpress

Product Code. 16461686
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461686 500 mg 500mg
16441096 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461686 Supplier Medchem Express Supplier No. HY13669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lomustine (CCNU; NSC 79037) is a DNA alkylating agent, with antitumor activity.

Lomustine (CCNU

Specifications

Chemical Name or Material Lomustine
Physical Form Solid
CAS 13010-47-4
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C9H16ClN3O2
For Use With (Application) Cancer-programmed cell death
Synonym CCNU NSC 79037
Solubility Information DMSO : ≥ 100 mg/mL (427.90 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1CCCCC1)N(CCCl)N=O
Molecular Weight (g/mol) 233.7
Formula Weight 233.7
Percent Purity 98.56%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.