missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lomustine

Product Code. 16627032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16627032 250 mg 250mg
16617032 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16627032 Supplier Thermo Scientific Chemicals Supplier No. 465492500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13010-47-4 | C9H16ClN3O2 | 233.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13010-47-4
Molecular Formula C9H16ClN3O2
Molecular Weight (g/mol) 233.70
MDL Number MFCD00012392
InChI Key GQYIWUVLTXOXAJ-UHFFFAOYSA-N
IUPAC Name 3-(2-chloroethyl)-1-cyclohexyl-3-nitrosourea
SMILES ClCCN(N=O)C(=O)NC1CCCCC1

Specifications

Alpha Vector LOMUSTINE
Melting Point 89°C to 93°C
Color Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 250 mg
UN Number 2811
Sensitivity Light sensitive
Solubility Information soluble aq. soln., soluble in acetone, chloroform, ethanol
Formula Weight 233.7
Physical Form Powder
Chemical Name or Material Lomustine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lomustine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
Hazard Statement
  • H301-Toxic if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Lomustine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.