missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lomitapide mesylate
GREENER_CHOICE

Product Code. 18786426
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18786426

Brand: Tocris Bioscience™ 7492/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent microsomal triglyceride transfer protein (MTP) inhibitor

Lomitapide mesylate is a potent inhibitor of microsomal triglyceride transfer protein (IC50 = 8 nM). Lomitapide inhibits or normalizes lipoprotein production in rodents and in a rabbit model of hypercholesterolemia. Lomitapide is antimalarial; it inhibits hemozoin formation and growth of CQ-sensitive (PfNF54) and multidrug-resistant (PfK1) strains of P. falciparum (IC50 values are 0.56 μM and 0.69 μM respectively). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2,2,2-Trifluoroethyl)-9-[4-[4-[[[4 '-(trifluoromethyl)[1,1 '-biphenyl]-2-yl]carbonyl]amino]-1-piperidinyl]butyl]-9 H-fluorene-9-carboxamide methanesulfonate
CAS 2059395-52-5
Quantity 50 mg
Target Lipid Metabolism Inhibitors
Molecular Formula C39H37F6N3O2.CH3SO3H
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.