missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Lomitapide mesylate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Lomitapide mesylate is a potent inhibitor of microsomal triglyceride transfer protein (IC50 = 8 nM). Lomitapide inhibits or normalizes lipoprotein production in rodents and in a rabbit model of hypercholesterolemia. Lomitapide is antimalarial; it inhibits hemozoin formation and growth of CQ-sensitive (PfNF54) and multidrug-resistant (PfK1) strains of P. falciparum (IC50 values are 0.56 μM and 0.69 μM respectively). Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-(2,2,2-Trifluoroethyl)-9-[4-[4-[[[4 '-(trifluoromethyl)[1,1 '-biphenyl]-2-yl]carbonyl]amino]-1-piperidinyl]butyl]-9 H-fluorene-9-carboxamide methanesulfonate |
| CAS | 2059395-52-5 |
| Quantity | 10 mg |
| Target | Lipid Metabolism Inhibitors |
| Molecular Formula | C39H37F6N3O2.CH3SO3H |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction