missing translation for 'onlineSavingsMsg'
Learn More

Lomitapide, MedChemExpress

Product Code. 16446128
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446128 10 mg 10mg
16422267 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446128 Supplier Medchem Express Supplier No. HY14667
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lomitapide (AEGR-733; BMS-201038) is a potent inhibitor of microsomal triglyceride-transfer protein (MTP) with an IC50 of 8 nM in vitro.

Lomitapide (AEGR-733

Specifications

Chemical Name or Material Lomitapide
CAS 182431-12-5
Color White
Purity Grade Notes Research
Molecular Formula C39H37F6N3O2
Quantity 10 mg
Synonym AEGR-733 BMS-201038
Solubility Information DMSO : ≥ 100 mg/mL (144.15 mM)
SMILES O=C(C1(CCCCN2CCC(NC(C3=CC=CC=C3C4=CC=C(C(F)(F)F)C=C4)=O)CC2)C5=C(C6=C1C=CC=C6)C=CC=C5)NCC(F)(F)F
Molecular Weight (g/mol) 693.72
Formula Weight 693.72
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.