missing translation for 'onlineSavingsMsg'
Learn More

Lomibuvir, MedChemExpress

Product Code. p-200101789
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488198

Brand: Medchem Express HY75800

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lomibuvir (VX-222), a selective, non-nucleoside polymerase inhibitor, targets thumb pocket 2 of the HCV NS5B polymerase (RdRp) with a Kd of 17 nM. Lomibuvir inhibits the 1b/Con1 HCV subgenomic replicon with an EC50 of 5.2 nM. Lomibuvir preferentially inhibits elongative RNA synthesis rather than de novo-initiated RNA synthesis.

Lomibuvir (VX-222), a selective, non-nucleoside polymerase inhibitor, targets thumb pocket 2 of the HCV NS5B polymerase (RdRp) with a Kd of 17 nM. Lomibuvir inhibits the 1b/Con1 HCV subgenomic replicon with an EC50 of 5.2 nM. Lomibuvir preferentially inhibits elongative RNA synthesis rather than de novo-initiated RNA synthesis.

Specifications

Chemical Name or Material Lomibuvir
CAS 1026785-55-6
Color White
Purity Grade Notes Research
Molecular Formula C25H35NO4S
Quantity 10 mg
Synonym VX-222
Solubility Information DMSO : 100 mg/mL (224.41 mM; Need ultrasonic)
SMILES O=C(C1=C(N([C@@H]2CC[C@@H](O)CC2)C([C@H]3CC[C@H](C)CC3)=O)C=C(C#CC(C)(C)C)S1)O
Molecular Weight (g/mol) 445.61
Formula Weight 445.61
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.