missing translation for 'onlineSavingsMsg'
Learn More

Lomeguatrib, MedChemExpress

Product Code. 16485798
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16485798

Brand: Medchem Express HY13668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lomeguatrib is a O6-methylguanine-DNA methyltransferase (MGMT) inhibitor, with IC50s of 9 nM in cell-free assay and ∼6 nM in MCF-7 cells.

Lomeguatrib is a O6-methylguanine-DNA methyltransferase (MGMT) inhibitor, with IC50s of 9 nM in cell-free assay and ∼6 nM in MCF-7 cells.

Specifications

Chemical Name or Material Lomeguatrib
Physical Form Solid
CAS 192441-08-0
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H8BrN5OS
For Use With (Application) Cancer-programmed cell death
Synonym PaTrin-2
Solubility Information DMSO : ≥ 56 mg/mL (171.69 mM)
SMILES NC1=NC(OCC2=CC(Br)=CS2)=C3N=CNC3=N1
Molecular Weight (g/mol) 326.17
Formula Weight 326.17
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.