missing translation for 'onlineSavingsMsg'
Learn More

Lomefloxacin Hydrochloride, TRC

Product Code. 30134610
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134610 50 mg 50mg
30005361 1 g 1g
30010705 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134610 Supplier Toronto Research Chemicals Supplier No. TRCL46941550MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98079-52-8
Molecular Formula C17 H19 F2 N3 O3 . Cl H
Molecular Weight (g/mol) 387.81
Synonym Lomefloxacin Hydrochloride, 1-Ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxo-quinoline-3-carboxylic acid hydrochloride, 3-Quinolinecarboxylic acid, 1-ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, hydrochloride (1:1), 3-Quinolinecarboxylic acid, 1-ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, monohydrochloride (9CI), Alkaflox, Bareon, Chimono, Lomebact, Lomef 400, Lomflox, Lomitas, Lomoxin, Maxaquin, Mazaquin, NY 198, Orchacin, SC 47111, Uniquin
IUPAC Name 1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMILES Cl.CCN1C=C(C(=O)O)C(=O)c2cc(F)c(N3CCNC(C)C3)c(F)c12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H19F2N3O3.ClH/c1-3-21-8-11(17(24)25)16(23)10-6-12(18)15(13(19)14(10)21)22-5-4-20-9(2)7-22;/h6,8-9,20H,3-5,7H2,1-2H3,(H,24,25);1H
Formula Weight 387.1161
Chemical Name or Material Lomefloxacin hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.