missing translation for 'onlineSavingsMsg'
Learn More

LOK Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-99860PCP

Product Code. 18619096

  • £309.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LOK Polyclonal antibody specifically detects LOK in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LOK
Polyclonal
Antigen and protein A Affinity-purified
Store at 4C in the dark.
ELISA
PerCP
Protein Kinase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.