missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOH12CR1 Antibody (OTI1E1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72518AF750
This item is not returnable.
View return policy
Description
LOH12CR1 Monoclonal specifically detects LOH12CR1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| LOH12CR1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BORCS5 | |
| Full length recombinant protein of human LOH12CR1 (NP_477517) produced in HEK293T cells. | |
| 0.1 mL | |
| 118426.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1E1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| LOH1CR12, loss of heterozygosity 12 chromosomal region 1 protein, loss of heterozygosity, 12, chromosomal region 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction