missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LogIt™ USB PC Adapter Set
Click to view available options
For Use With (Application):
Long Range Wireless Logging
Description
Contains Air Link data logger adapter and USB PC adapters for long range Bluetooth wireless logging LogIT
Specifications
Specifications
| Type | Adapter Set, Bluetooth |
| For Use With (Application) | Long Range Wireless Logging |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction