Learn More
LogIt™ Motion and Distance Sensor
Specifications
Specifications
| Type | Sensor, Motion and Distance |
| Includes | Cable and Instructions |
| For Use With (Application) | Displaying and investigating Speed, Distance, and Time relationships using trolleys, model cars, or people. |