Learn More
LogIt™ Infrared Sensor
Click to view available options
Range:
0 to 1820 W/m2
Description
Measures radiated energy and temperature, 250 to 1,800W per meter squared with comprehensive investigation guide LogIT
Specifications
Specifications
| Type | Sensor Microsense Infrared |
| For Use With (Application) | Measures Radiated Energy and Temperature |
| Range | 0 to 1820 W/m2 |