missing translation for 'onlineSavingsMsg'
Learn More

Loganin, MedChemExpress

Product Code. 16469096
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469096 100 mg 100mg
16454847 50 mg 50mg
16419008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16469096 Supplier Medchem Express Supplier No. HYN0512
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Loganin, a major iridoid glycoside obtained from Corni fructus, has been shown to have anti-inflammatory and anti-shock effects.

Loganin, a major iridoid glycoside obtained from Corni fructus, has been shown to have anti-inflammatory and anti-shock effects.

Specifications

Chemical Name or Material Loganin
CAS 18524-94-2
Color White
Purity Grade Notes Research
Molecular Formula C17H26O10
Quantity 100 mg
Synonym Loganoside
Solubility Information DMSO : 50 mg/mL (128.08 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO)([H])O[C@H]2[C@@]([C@H]3C)([H])[C@@](C[C@@H]3O)([H])C(C(OC)=O)=CO2
Molecular Weight (g/mol) 390.38
Formula Weight 390.38
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.