missing translation for 'onlineSavingsMsg'
Learn More

Lofexidine hydrochloride, 98+%

Product Code. 15456109
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456109 5 g 5g
15446109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456109 Supplier Thermo Scientific Alfa Aesar Supplier No. J63960.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alpha-2 adrenergic agonist | CAS: 21498-08-8 | C11H13Cl3N2O | 295.588 g/mol

Alpha-2 adrenergic agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21498-08-8
Molecular Formula C11H13Cl3N2O
Molecular Weight (g/mol) 295.588
MDL Number MFCD00917022
InChI Key DWWHMKBNNNZGHF-UHFFFAOYSA-N
Synonym lofexidine hydrochloride, lofexidine hcl, lofetensin, loxacor, lofexidine.hcl, loxacor hydrochloride, lofetensin hydrochloride, 2-1-2,6-dichlorophenoxy ethyl-4,5-dihydro-1h-imidazole hydrochloride, lofexidine hydrochloride usan
PubChem CID 30667
IUPAC Name 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride
SMILES CC(C1=NCCN1)OC2=C(C=CC=C2Cl)Cl.Cl

Specifications

Melting Point 225°C to 232°C
Color Yellow
Quantity 5 g
UN Number UN2811
Merck Index 14,5558
Formula Weight 295.6
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Lofexidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.