missing translation for 'onlineSavingsMsg'
Learn More

Lodoxamide, MedChemExpress

Product Code. 16486426
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486426

Brand: Medchem Express HY14270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lodoxamide (U-42585E free acid) is an antiallergic compound acting as a mast-cell stabilizer for the treatment of asthma and allergic conjunctivitis.

Lodoxamide (U-42585E free acid) is an antiallergic compound acting as a mast-cell stabilizer for the treatment of asthma and allergic conjunctivitis.

Specifications

Chemical Name or Material Lodoxamide
Physical Form Solid
CAS 53882-12-5
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H6ClN3O6
For Use With (Application) COVID-19-immunoregulation
Synonym U-42585E free acid
Solubility Information DMSO : 50 mg/mL (160.45 mM; Need ultrasonic)
SMILES OC(C(NC1=C(Cl)C(NC(C(O)=O)=O)=CC(C#N)=C1)=O)=O
Molecular Weight (g/mol) 311.63
Formula Weight 311.63
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.