missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Locust Bean Gum (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Locust Bean Gum |
| CAS | 9000-40-2 |
| Molecular Formula | C10H11ClN2O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H11ClN2O2/c1-2-15-10(14)9(11)13-12-8-6-4-3-5-7-8/h3-7,12H,2H2,1H3/b13-9+ |
| SMILES | O[C@H]1[C@H](OC)[C@@H](CO)O[C@@H](O[C@H]2[C@H](O)[C@H](O)[C@H](O[C@H]3[C@H](O)[C@H](O)[C@H](O[C@H]4[C@H](O)[C@H](O)[C@H](OC)O[C@@H]4CO)O[C@@H]3CO[C@@H]5[C@H](O)[C@@H](O)[C@@H](O)[C@@H](CO)O5)O[C@@H]2CO)[C@H]1O |
| Molecular Weight (g/mol) | 226.66 |
| Formula Weight | 856.306 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?