missing translation for 'onlineSavingsMsg'
Learn More

Locust Bean Gum (Technical Grade), TRC

Product Code. 30479398
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479398 1 g 1g
30481437 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30479398 Supplier Toronto Research Chemicals Supplier No. TRCL4729251G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-40-2
Molecular Formula C10H11ClN2O2
Molecular Weight (g/mol) 226.66
SMILES O[C@H]1[C@H](OC)[C@@H](CO)O[C@@H](O[C@H]2[C@H](O)[C@H](O)[C@H](O[C@H]3[C@H](O)[C@H](O)[C@H](O[C@H]4[C@H](O)[C@H](O)[C@H](OC)O[C@@H]4CO)O[C@@H]3CO[C@@H]5[C@H](O)[C@@H](O)[C@@H](O)[C@@H](CO)O5)O[C@@H]2CO)[C@H]1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H11ClN2O2/c1-2-15-10(14)9(11)13-12-8-6-4-3-5-7-8/h3-7,12H,2H2,1H3/b13-9+
Formula Weight 856.306
Chemical Name or Material Locust Bean Gum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.