missing translation for 'onlineSavingsMsg'
Learn More

Locostatin

Product Code. 15466379
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466379 10 mg 10mg
15476379 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466379 Supplier Thermo Scientific Alfa Aesar Supplier No. J64425.MA
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 133812-16-5 | C14H15NO3 | 245.278 g/mol

Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Locostatin is a cell-permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133812-16-5
Molecular Formula C14H15NO3
Molecular Weight (g/mol) 245.278
MDL Number MFCD00278769
InChI Key UTZAFVPPWUIPBH-QSLRECBCSA-N
Synonym locostatin, s-4-benzyl-3-but-2-enoyl oxazolidin-2-one, s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one, 4s-n-crotonyl-4-benzyl-2-oxazolidinone, 4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone, 4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one, cell sheet migration inhibitor, locostatin, 4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one, locostatin hplc, 3-crotonyl-4beta-benzyloxazolidine-2-one
PubChem CID 5702600
IUPAC Name (4S)-4-benzyl-3-[(E)-but-2-enoyl]-1,3-oxazolidin-2-one
SMILES CC=CC(=O)N1C(COC1=O)CC2=CC=CC=C2

Specifications

Color White
Quantity 10 mg
Formula Weight 245.27
Physical Form Powder
Chemical Name or Material Locostatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.