Learn More
JULABO™ Lockable Bath Cover
The lockable bath cover minimizes oil vapor and odor formation at high bath temperatures.
Brand: JULABO™ 9970243
This item is not returnable.
View return policy
Description
- Lockable bath cover for bath BC12/600F/601F/1000F/1000FW
Specifications
Circulators | |
Variable | |
Bath Cover | |
None |