missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LOC81691 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559283
This item is not returnable.
View return policy
Description
Immunogen sequence: KLKFLAKVIL GKDIQCPDRL GHDATEDART ILELARYFLK HGPKKIAELN LEALANHQEI QAAGQEPKNT AEVLQHPNTS VLECLDSVGQ KLLFLTRETD AGELPSSRNC QTIKCLS Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 67%.
REXO5 (RNA Exonuclease 5) is a Protein Coding gene. Among its related pathways are Ribosome biogenesis in eukaryotes. An important paralog of this gene is REXO1.
Specifications
| LOC81691 | |
| Polyclonal | |
| Unconjugated | |
| REXO5 | |
| 44M2.3; Exonuclease NEF-sp; LOC81691; putative RNA exonuclease NEF-sp; REXO5; RNA exonuclease 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 81691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96IC2 | |
| REXO5 | |
| Recombinant protein corresponding to Human LOC81691. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction