missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LOC14
High affinity protein disulfide isomerase (PDI) modulator
Specifications
| Chemical Name or Material | 2-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]methyl]-1,2-benzisothiazol-3(2H)-one |
|---|---|
| CAS | 877963-94-5 |
Description
High affinity protein disulfide isomerase (PDI) modulator (Kd = 62 nM). Forces PDI to adopt an oxidized conformation and suppresses its activity. Neuroprotective.Specifications
| 2-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]methyl]-1,2-benzisothiazol-3(2H)-one | |
| 877963-94-5 |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title