missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ LOC124220 293T Cell Transient Overexpression Lysate (Denatured)

Product Code. 16187573
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 16187573

Brand: Abnova™ H00124220T01.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Denatured cell transient overexpression lysate of LOC124220 293T.

  • Transient overexpression cell lysate was tested with Anti-LOC124220 antibody (H00124220-B01) by Western Blots.

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description Gene ID: 124220
Gene Accession No. ENSP00000371715
Plasmid pCMV-LOC124220 full-length
Quality Control Testing Transient overexpression cell lysate was tested with Anti-LOC124220 antibody by Western Blots
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Product Type Similar to common salivary protein 1
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.