missing translation for 'onlineSavingsMsg'
Learn More

Lobetyolin, MedChemExpress

Product Code. 16491378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491378 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491378 Supplier Medchem Express Supplier No. HYN0327
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lobetyolin, a bioactive compound, is derived from Codonopsis pilosula. Lobetyolin has anti-inflammatory, anti-oxidative and xanthine oxidase inhibiting activities. Lobetyolin also induces the apoptosis via the inhibition of ASCT2-mediated glutamine metabolism.

Lobetyolin, a bioactive compound, is derived from Codonopsis pilosula. Lobetyolin has anti-inflammatory, anti-oxidative and xanthine oxidase inhibiting activities. Lobetyolin also induces the apoptosis via the inhibition of ASCT2-mediated glutamine metabolism.

Specifications

Chemical Name or Material Lobetyolin
CAS 129277-38-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H28O8
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (126.13 mM; Need ultrasonic) ∣H2O : 50 mg/mL (126.13 mM; Need ultrasonic)
SMILES O[C@H](C#CC#C/C=C/C)[C@@H](/C=C/CCCO)O[C@@H]([C@@H]([C@H]1O)O)O[C@@H]([C@H]1O)CO
Molecular Weight (g/mol) 396.43
Formula Weight 396.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.