missing translation for 'onlineSavingsMsg'
Learn More

Lobeline sulfate, 99%

Product Code. 11413567
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413567 1 g 1g
11423567 5 g 5g
11433567 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413567 Supplier Thermo Scientific Alfa Aesar Supplier No. A17326.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 134-64-5 | C44H56N2O8S | 773.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-64-5
Molecular Formula C44H56N2O8S
Molecular Weight (g/mol) 773.00
MDL Number MFCD00135592
InChI Key GRZMOSSVIPFGFF-GNJLJDPWSA-N
Synonym lobeline sulfate, bantron, lobelin sulfate, lobeline sulphate, unii-4cj480v2hp, lobeline, sulfate 2:1 salt, ethanone, 2-6-2-hydroxy-2-phenylethyl-1-methyl-2-piperidinyl-1-phenyl-, 2theta-2alpha,6alpha s-, sulfate 2:1 salt, bis--lobeline ; sulfuric acid, smokeless tn, 2-6-2-hydroxy-2-phenylethyl-1-methylpiperidin-2-yl-1-phenylethanone sulfate
PubChem CID 120636
IUPAC Name 2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;sulfuric acid
SMILES OS(O)(=O)=O.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1

Specifications

Quantity 1 g
UN Number UN1544
Formula Weight 773.01
Percent Purity 99%
Chemical Name or Material Lobeline sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.