missing translation for 'onlineSavingsMsg'
Learn More

Loading Control (Simple Western) Antibody Pack, Novus Biologicals™

Product Code. 18372033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
4 Vials
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372033 4 Vials Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372033 Supplier Novus Biologicals Supplier No. NBP306859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Loading Control (Simple Western) Monoclonal antibody specifically detects Loading Control (Simple Western) in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Loading Control (Simple Western)
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Gene Alias GAPDH, Hsp60
Host Species Mouse
Quantity 4 Vials
Regulatory Status RUO
Research Discipline Loading Controls
Target Species Human, Mouse, Rat
Content And Storage Store at 4°CC.
Product Type Antibody Packs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.