missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Loading Control (Simple Western) Polyclonal antibody specifically detects Loading Control (Simple Western) in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot
Specifications
Specifications
| Antigen | Loading Control (Simple Western) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Gene Alias | GAPDH, Hsp60 |
| Host Species | Rabbit |
| Quantity | 4 Vials |
| Regulatory Status | RUO |
| Research Discipline | Loading Controls |
| Target Species | Human, Mouse, Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?