missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
GREENER_CHOICE

Product Code. 17812998
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +330°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
17812998 -10°C to +330°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17812998 Supplier Brannan™ Supplier No. 44/814/0 + 49/503/0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LO-tox™ lab thermometer complete with calibration certificate.

  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK

Specifications

Calibration Points 0°C, 100°C, 250°C
Certifications/Compliance ISO 17025 UKAS
Immersion Type Partial Immersion
Length (Metric) 30.5 cm
Material Glass
Thermometer Fill Brown Spirit
Max. Temperature (English) 626°F
Max. Temperature (Metric) +330°C
Min. Temperature (English) 14°F
Min. Temperature (Metric) -10°C
Temperature Range (English) 14°F to 626°F
Temperature Range (Metric) -10°C to +330°C
Accuracy ± 2.0°C at 90°C
Subdivision 2°C
Immersion Length 76 mm
Width (Metric) 1.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.