missing translation for 'onlineSavingsMsg'
Learn More

LNX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18347864 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347864 100 μL 100µL
18351496 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18347864 Supplier Novus Biologicals Supplier No. NBP304570100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LNX2 Polyclonal specifically detects LNX2 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LNX2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FLJ23932, FLJ38000, ligand of Numb protein X 2, ligand of numb-protein X 2, MGC46315, Numb-binding protein 2, PDZ domain containing ring finger 1, PDZ domain-containing RING finger protein 1, PDZRN1FLJ12933
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human LNX2 (NP_699202.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 222484
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.